Learn More
MilliporeSigma™ Supelco™ PEEK Inline Microfilter
Install between 2 pieces of capillary tubing, using Microtight tubing sleeves. PEEK housing with special PEEK fitting that encases the 0.5 μm PEEK frit. Through hole 0.006 in., swept volume 0.24 μL. Includes 2 MicroFingertight nuts, and 5 frits (1 installed).
Supplier:  MilliporeSigma™ Supelco™ 502707
Catalog No. 11-101-0425
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Liquid Chromatography
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.