Learn More
Cytiva PEEK Tubing 1/16 Inch 0.75mm ID
Supplier:  Cytiva 18111253
Encompass
Tubing (PEEK) for use with KTA systems. Green color for easy identification. 2 m length with ID 0.75mm and OD 1/16 in.0 Pack of 1.
Catalog No. 50-105-5648