Learn More
Cytiva 2 m beige PEEK tubing with 1.0 mm internal diameter for use with ÄKTA start, ÄKTA go, ÄKTA pure, and ÄKTA avant
Catalog No. 502389576 Shop All Cytiva Products
Encompass
2 m beige PEEK tubing with 1.0 mm internal diameter for use with ÄKTA start, ÄKTA go, ÄKTA pure, and ÄKTA avant
Catalog No. 50-238-9576 Supplier Cytiva Supplier No. 18111583
2 m beige PEEK tubing with 1.0 mm internal diameter for use with ÄKTA start, ÄKTA go, ÄKTA pure, and ÄKTA avant