Learn More
Molecular Dimensions PEG 4000
SDP
Supplier:  Molecular Dimensions MD225011
Encompass
Protein Crystallography. 250mL bottle of 50% w/v PEG 4000 (Poly(ethylene glcyol) 4000) solution. Made with ultra-purified water.
Catalog No. NC0362386
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.