Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEG Monoclonal Antibody (5E10E9), THE™, GenScript™

Catalog No. p-7222685
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0587 100 μg
50-212-0588 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-212-0587 Supplier Genscript Corporation Supplier No. A01795100
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. For long-term storage, adding glycerol to a final concentration of 50% (v/v) in the reconstituted antibody solution is recommended. PEG (Polyethylene glycol) is a polyether compound with many applications from industrial manufacturing to medicine. PEGylation is a technology that covalent couples non-toxic, hydrophilic polyethylene glycol (PEG) to the drug. It is an FDA-approved method for the delivery of protein drugs.PEG modification can reduce the drug immunogenicity and antigenicity. PEGylated drug decelerates renal excretion, improves stability towards proteolysis and increases its half life in blood. Accurate and sensitive quantification of PEG conjugates is important for PEG conjugated product development and pharmaceutical study. Polyethylene glycol (PEG) antibody is a useful tool for the detection of PEGylated molecules.GenScript THE™ PEG Antibody, mAb, Mouse is produced from the hybridoma resulting from fusion of Sp2/0 myeloma and lymphocytes obtained from mouse immunized with PEG conjugated to KLH.

PEG Monoclonal antibody specifically detects PEG in Chemical samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PEG
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5E10E9
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias Poly Ethylene Glycol; Polyethylene Glycol
Host Species Mouse
Immunogen PEG conjugated to KLH
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgM
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.