Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ HeadsUp™ Lites Headlamps

Catalog No. 19130092 Shop All Pelican Products Inc. Products
Encompass_Preferred
Change view
Click to view available options
Battery Run Time:
10 hr.
19 hr.
2.15 hr.
Includes:
3 AAA alkaline batteries
3 AAA alkaline batteries, adjustable cloth strap and rubber strap
4 AA alkaline batteries
Length (Metric):
3.8 cm
5.5 cm
11.8 cm
Lumen:
26 lm
30 lm
79 lm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Run Time Includes Length (Metric) Lumen
19-130-092 2.15 hr. 3 AAA alkaline batteries 5.5 cm 26 lm
19-130-091 19 hr. 3 AAA alkaline batteries, adjustable cloth strap and rubber strap 3.8 cm 30 lm
19-501-202 10 hr. 4 AA alkaline batteries 11.8 cm 79 lm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-130-092 Supplier Pelican™ Supplier No. 2620
This item is not returnable. View return policy

Includes

2610 and 2620 Models include three AAA batteries; 2680 Model supplied with four AA batteries.

Hands-free light when and where you need it

  • Adjustable cloth strap fits comfortably and securely on head
  • Rubber strap grips tightly to hardhats and helmets

2610 Model

  • 47-hour burn time on low; 19 hours on high
  • Dual-mode button activates one or all three LEDs for full brightness
  • Length: 1.5 in. (3.8cm)
  • Weight (with batteries): 3.2 oz. (90.1g)
  • Powered by three AAA alkaline cells (included)

2620 Model

  • 18-hour burn time on low; 2.25 hours on high
  • Hybrid dual lamps; Xenon for intense bright light or three LEDs
  • Multi-angle pivoting head allows light to be aimed where needed
  • Length: 2.2 in. (5.5cm)
  • Weight (with batteries): 4.8 oz. (136kg)
  • Powered by three AAA alkaline cells (included)

2680 Model

  • Outfitted with Recoil LED module
  • Uses a 1 watt LED aimed backwards into the reflector
  • 100% of the light is captured and aimed forward as a tight collimated beam as bright as an incandescent
  • Body is made of tough, corrosion proof, submersible, and virtually indestructible ABS resin
  • Powered by four AA alkaline cells (included)
  • Comes with a rubber strap for hard hat use and an adjustable cloth strap for bare head

2690 Model

  • Impact and chemical-resistant ABS'water resistant for all-weather use
  • Provides 60 lumens of LED light
  • 10 hours of battery burn time
  • With rotary bezel switch
  • Adjustable cloth strap; heavy-duty rubber strap
  • Powered by three AAA alkaline cells (included)

Certifications

2610, 2680, and 2690 Models are UL listed for certain Class I, Division 1 hazardous locations; CE marked. 2680 Model is UL listed for certain Class I, Division 2 hazardous locations; CE marked.

Specifications

Gas Fill Xenon
Voltage 4.5 V
Weight (English) 4.8 oz.
Weight (Metric) 0.14 kg
Battery Run Time 2.15 hr.
Battery Type Alkaline
Certifications/Compliance UL, Class I, Division 2, CE
Color Black
For Use With (Application) Hands free light when and where you need it, adjustable cloth strap fits comfortably and securely on head
Includes 3 AAA alkaline batteries
Length (English) 2.16 in.
Length (Metric) 5.5 cm
Lumen 26 lm
Material ABS Resin
Warranty Lifetime
Product Type Headlamp
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.