Learn More
Description
Includes
2610 and 2620 Models include three AAA batteries; 2680 Model supplied with four AA batteries.
- Adjustable cloth strap fits comfortably and securely on head
- Rubber strap grips tightly to hardhats and helmets
2610 Model
- 47-hour burn time on low; 19 hours on high
- Dual-mode button activates one or all three LEDs for full brightness
- Length: 1.5 in. (3.8cm)
- Weight (with batteries): 3.2 oz. (90.1g)
- Powered by three AAA alkaline cells (included)
2620 Model
- 18-hour burn time on low; 2.25 hours on high
- Hybrid dual lamps; Xenon for intense bright light or three LEDs
- Multi-angle pivoting head allows light to be aimed where needed
- Length: 2.2 in. (5.5cm)
- Weight (with batteries): 4.8 oz. (136kg)
- Powered by three AAA alkaline cells (included)
2680 Model
- Outfitted with Recoil LED module
- Uses a 1 watt LED aimed backwards into the reflector
- 100% of the light is captured and aimed forward as a tight collimated beam as bright as an incandescent
- Body is made of tough, corrosion proof, submersible, and virtually indestructible ABS resin
- Powered by four AA alkaline cells (included)
- Comes with a rubber strap for hard hat use and an adjustable cloth strap for bare head
2690 Model
- Impact and chemical-resistant ABS'water resistant for all-weather use
- Provides 60 lumens of LED light
- 10 hours of battery burn time
- With rotary bezel switch
- Adjustable cloth strap; heavy-duty rubber strap
- Powered by three AAA alkaline cells (included)
Certifications
2610, 2680, and 2690 Models are UL listed for certain Class I, Division 1 hazardous locations; CE marked. 2680 Model is UL listed for certain Class I, Division 2 hazardous locations; CE marked.
Specifications
Specifications
| Light Source | Recoil LED™ |
| Voltage | 6 V |
| Weight (English) | 9.7 oz. |
| Weight (Metric) | 0.27 kg |
| Battery Run Time | 10 hr. |
| Battery Type | Alkaline |
| Certifications/Compliance | UL, Class I, Division 2, CE |
| Color | Black |
| For Use With (Application) | Hands free light when and where you need it, adjustable cloth strap fits comfortably and securely on head |
| Includes | 4 AA alkaline batteries |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.