Learn More
Agilent BioTek Peltier Cooling Module
GSA_VAGREENER_CHOICE
Catalog No. BT1700001 Shop All Agilent Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Cytation 5 and Cytation 1
Catalog No. For Use With (Equipment)
BT1700001 Cytation 5 and Cytation 1
1 options
Catalog No. BT1700001 Supplier Agilent BioTek Supplier No. 1700001S
Add to Cart
Add to Cart
Request a Quote
Peltier Cooling Module helps maintain internal environmental stability and promotes rapid cool-down.
  • The Peltier Cooling Module promotes a rapid interior cool down after incubated processes, to allow efficient switching between multiple applications without unwanted temperature influences
  • For temperature sensitive assays typically run at ambient, or room temperature, the Cooling Module maintains environmental stability within Cytation, allowing 1 degree celcius rise in ambient temperature, regardless of external and internal temperature fluctuation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cytation 5 and Cytation 1
For Use With (Application) ELISA, Fluorescence ELISA, Protein Quantification, Nucleic Acid Quantification