Learn More
Perkin Elmer US LLC Peltier™ Thermostatted Single Sample Holder
Supplier:  Perkin Elmer US LLC B2000159
Encompass_Preferred
This accessory is compatible with the LAMBDA 25/35/45 spectrometers. It requires, but does not include, the Acccessory Board (L6000500).
Catalog No. 50-210-4402
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.