Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ Earmuffs, Green

Catalog No. 17000707 Shop All 3M Products
Change view
Click to view available options
Model:
X1A
X1P3E
X1P51E
X1P5E
CSA Z94.2:
Class A
Class B
Dielectric:
No
Yes
Style:
Hard Hat
Over-the-head
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model CSA Z94.2 Dielectric Style
17-000-707 X1P51E Class B Yes Hard Hat
17-000-631 X1A Class A Yes Over-the-head
17-000-636 X1P3E Class B No Hard Hat
19-153-673 X1P5E Class B Yes Hard Hat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-000-707 Supplier 3M™ Supplier No. 7100135816

3M™ PELTOR™ Earmuffs X1 are our lightweight hearing protectors with comfortable, soft ear cushions.

  • Compatability with full brim hard hats
  • Protects against arc flash and other electrical hazards with the electrically insulated (dielectric) attachment arms
  • Ensures prolonged, consitant protection resulting from durable attachment arms
  • Provides high level of protection relative to earcup depth
  • Low-profile cup design for improved compatibility with other personal protection equipment (PPE)

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black, Green
ANSI S3.1974 Yes
Cushion Material Foam
CSA Z94.2 Class B
Cushion Color Black
For Use With (Equipment) Fits a Wide Variety of Industrial Full Brim Style Hard Hats
For Use With (Application) Utilities, Mining, Oil and Gas, Electrical Contractors
Model X1P51E
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 20 dB
Product Type X1 Series Ear Muffs
Dielectric Yes
Material ABS, Polyurethane
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.