Learn More
Description
- FLX2 Cables for use with PELTOR™ CH-3 Headset with push to talk (PTT) button
- Coiled polyurethane cable
- Aramid fiber reinforced
- Flexible and adaptable
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | Kenwood Two-Pin |
| For Use With (Application) | For Use with CH-3 Headsets with PTT Button |
| Model | FLX2-36 |
| Pairs per Package | 1 Ea. |
| Product Type | FLX2 Cable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.