Learn More
Description
- For use with CH-3 Headset with external PTT Button
- J11 Plug Type
- Coiled polyurethane cable
- Aramid fiber reinforced
- Robust and highly flexible
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | For use with CH-3 Headsets with external PTT Button |
| For Use With (Application) | Enables connection to two-way communication for CH-3 Headsets |
| Model | FLX2-201 |
| Pairs per Package | 1 Ea. |
| Product Type | FLX2 Airline Cable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.