Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ WS™ LiteCom PRO III Headset

Catalog No. 19072179 Shop All 3M Products
Change view
Click to view available options
Includes:
Hard Hat Attached with Bluetooth™
Hard Hat Attached with Bluetooth™ and a jack to connect to external devices
Headband with Bluetooth™
Headband with Bluetooth™ and a jack to connect to external devices
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-072-179 Headband with Bluetooth™
19-072-177 Headband with Bluetooth™ and a jack to connect to external devices
19-072-178 Hard Hat Attached with Bluetooth™ and a jack to connect to external devices
19-072-180 Hard Hat Attached with Bluetooth™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-072-179 Supplier 3M™ Supplier No. 7100099585
Add to Cart
Add to Cart
Request a Quote

A hearing protector with built-in analog and digital programmable two-way communication radio with level-dependent hearing protection.

  • Built-in programmable two-way communication radio allows for clear, hands-free communication in high levels of noise without cumbersome cables or cords.
  • The headset can be programmed to use up to 70 unique two-way radio channels using analog and/or DMR digital modulation.
  • Simultaneous, seamless connection to multiple external devices using Bluetooth™ technology.
  • Noise attenuating ear cups help provide hearing protection in environments with potentially hazardous noise.
  • Environmental listening with level dependent hearing protection for face-to-face communication and situational awareness.
  • Noise-cancelling waterproof speech microphone with voice operated transmission (VOX) for hands-free operation; helps provide clear communication in noisy and demanding environments.
  • User-friendly controls with voice-guided menu.
  • Built tough for the working professional.
  • Headset is available in headband, neckband and hard hat attached configurations.
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Certifications/Compliance Certified by CSA Group as intrinsically Safe for Use in Hazardous Locations
Height (Metric) 224 mm
Model MT73H7F4D10NA-50
Weight (English) 3 lb.
Weight (Metric) 1.3 kg
Width (Metric) 155 mm
Noise Reduction Rating (NRR) 27 dB
Product Type Protective Communication
Style Foldable headband
Communication Details Environmental listening; External Audio Input; Bluetooth™ Technology
Battery Type Rechargable lithium ion pack
Battery Life 8 to 12 hours
Direction 2-way
Port Type 4-pin XLR
Series Lite-Com WS Series
Number of Channels Up to 70 unique channels 
Includes Headband with Bluetooth™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.