Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pemetrexed
GREENER_CHOICESDP

Catalog No. 618550
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-855-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 61-855-0 Supplier Tocris Bioscience™ Supplier No. 6185/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High affinity dihydrofolate reductase inhibitor; also inhibits thymidylate synthase, GARFT and AICART

High affinity dihydrofolate reductase inhibitor (Ki = 7 nM). Also inhibits thymidylate synthase, AICART and glycinamide ribonucleotide formyltransferase (Ki values are 109 nM, and 3.5 μM and 9.3 μM, respectively). Indirectly activates AMPK. Inhibits proliferation of cancer cell lines in vitro.

Chemical Identifiers

CAS 137281-23-3
Molecular Formula C20H21N5O6

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 427.41
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 427.417
Chemical Name or Material N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic acid
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.