Learn More
Selleck Chemical LLC Pemigatinib (INCB054828) 2MG 1513857-77-6 Pemazyre
Supplier: Selleck Chemical LLC S00882MG
Pemigatinib (INCB054828, Pemazyre) is an orally active and selective inhibitor of FGFR with IC50 of 0.4 nM, 0.5 nM, 1.2 nM and 30 nM for FGFR1, FGFR2, FGFR3 and FGFR4, respectively. Pemigatinib has the potential for cholangiocarcinoma. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.