Learn More
TARGETMOL CHEMICALS INC Pemirolast potassium 100MG
Supplier: TARGETMOL CHEMICALS INC T1425100MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Pemirolast Potassium is the potassium salt form of pemirolast, a pyrimidinone derivative with antiallergic property. Pemirolast potassium (TWT-8152) exerts its action by blocking the antigen-mediated calcium ion influx into mast cells. Purity 99.89%