Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Pendulum Investigation Kit
SDPSCI_ED

Catalog No. S83796 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S83796 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S83796 Supplier United Scientific™ Supplier No. PNLKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This unique supplies kit includes everything necessary for a full exploration of the basic features of pendulum motion in a complete but economical set.

Understanding pendulum motion is fundamental to the study of vibrations and waves. This unique kit includes everything necessary for a full exploration of the basic features of pendulum motion in a complete but economical set. The individual components are of proven design –the drilled balls have a larger hole on one end than the other to allow suspension at the center of mass, and the triple support features clamps that make adjusting the cord length precise and easy. Students can assess the principal factors of mass, length, and amplitude and even venture into a consideration of air resistance.

Contents:
  • 2 Drilled Balls (1 in., Steel)
  • 2 Drilled Balls (1 in., Aluminum)
  • 2 Drilled Balls (1 in., Wood)
  • 1 Pendulum Support with Three Positions and Clamp
  • 1 Half-meter Stick
  • 1 Support Base (6 x 9 in.), with 24 in. Support Rod
  • 1 Stopwatch
  • 1 Pendulum Cord Roll

Specifications

Grade 6 to 12
Age 12 to 19
For Use With (Application) Demonstrating Pendulums
Includes 3 drilled balls, pendulum support and clamp, half-meter stick, support base/rod, stopwatch, and pendulum cord.
Class Size 1 Lab Group
Product Type Physics Demonstration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.