Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin-Streptomycin soln. (100X), Thermo Scientific™

Catalog No. AAJ66092AM0
Change view
Click to view available options
Quantity:
10 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66092AM0 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66092AM0 Supplier Thermo Scientific Chemicals Supplier No. J66092AM0

An antibacterial solution composed of penicillin, a beta-lactam, and streptomycin, an aminoglycoside. A 100X solution

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Penicillin-Streptomycin soln is used as an antibacterial solution comprised of penicillin, a beta-lactam, and streptomycin, an aminoglycoside. A 100X solution. Penicillin (Penicillin G) works by inhibiting peptidoglycan synthesis, while Streptomycin inhibits protein synthesis.

Solubility
Fully miscible.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use. Contains penicillin (10,000 units/mL), a beta-lactam, and streptomycin (10,000 ug/mL), an aminoglycoside.
TRUSTED_SUSTAINABILITY

Specifications

Type Penicillin/Streptomycin
Chemical Name or Material Penicillin-Streptomycin
Physical Form Liquid
Color Colorless
Concentration 100X
Quantity 10 x 100 mL
MDL Number MFCD00216869
Synonym Pen-Strep
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.