Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Penicillium chrysogenum Derived from ATCC™ 10106™
SDP

Catalog No. 01259123 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Penicillium chrysogenum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
01-259-123 Penicillium chrysogenum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-123 Supplier Microbiologics™ Supplier No. 0178A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantitative microorganism preparation of Penicillium chrysogenum that delivers less than 100 CFU per 0.1 mL of hydrated suspension.

  • No dilutions required
  • Kit contains 5 pellets of a single enumerated strain and 5 vials of hydrating fluid (1.2 mL per vial)
  • Delivers 10 to 100 CFU per inoculum

Specifications

ATCC® ATCC 10106
Biosafety Level 1
Format Kit
Manufacturing Origin United States
Product Type Lyophilized Organism
For Use With (Application) Growth Promotion and Media Testing
Quantity 5 Vials
Species Penicillium chrysogenum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.