Learn More
Invitrogen™ PENK Polyclonal Antibody

Description
Positive Samples: Rat spinal cord, Rat brain.
Specifications
Specifications
| Antigen | PENK |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.02 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | PENK |
| Gene Accession No. | P01210, P04094, P22005 |
| Gene Alias | AI326464; ENK; enkephalin; enkephalin A; Leu-enkephalin; Met-enkephalin; Met-enkephalin-Arg-Gly-Leu; Met-enkephalin-Arg-Phe; Met-enkephalin-Arg-Ser-Leu; OGF; Opioid growth factor; PE; Penk; PENK(114-133); PENK(143-183); PENK(143-184); PENK(143-185); PENK(237-258); PENK(238-259); PENK(239-260); Penk1; Penk2; PENK-A; Penk-rs; PPA; preproenkephalin; preproenkephalin 1; preproenkephalin 2; proenkephalin; proenkephalin 1; proenkephalin 2; proenkephalin related sequence; proenkephalin-A; spermatogenic-specific proenkephalin; Synenkephalin |
| Gene Symbols | PENK |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.