Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PENK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA130928 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA130928 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA130928 Supplier Invitrogen™ Supplier No. PA130928
  • $580.00
    $478.65 / Each of 1

    Save $101.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

neuropeptide hormone; plays a role in the responses to stress and pain perception.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PENK
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PENK
Gene Accession No. P04094, P22005
Gene Alias AI326464; ENK; enkephalin; enkephalin A; Leu-enkephalin; Met-enkephalin; Met-enkephalin-Arg-Gly-Leu; Met-enkephalin-Arg-Phe; Met-enkephalin-Arg-Ser-Leu; OGF; Opioid growth factor; PE; Penk; PENK(114-133); PENK(143-183); PENK(143-184); PENK(143-185); PENK(237-258); PENK(238-259); PENK(239-260); Penk1; Penk2; PENK-A; Penk-rs; PPA; preproenkephalin; preproenkephalin 1; preproenkephalin 2; proenkephalin; proenkephalin 1; proenkephalin 2; proenkephalin related sequence; proenkephalin-A; spermatogenic-specific proenkephalin; Synenkephalin
Gene Symbols PENK
Host Species Rabbit
Immunogen CSYSKEVPEMEKRYGGFMRF Conjugated to KLH.
Purification Method Affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18619, 29237
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.