Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penta-O-acetyl-alpha-D-glucopyranose 97.0+%, TCI America™
SDP

Catalog No. G0225250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0225250G 250 g
G022525G 25 g
G022510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0225250G Supplier TCI America Supplier No. G0225250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 604-68-2
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
MDL Number MFCD00064071,MFCD00064081
InChI Key LPTITAGPBXDDGR-UHFFFAOYNA-N
Synonym alpha-d-glucose pentaacetate, a-d-glucopyranose pentaacetate, pentaacetyl-alpha-d-glucose, penta-o-acetyl-alpha-d-glucopyranose, unii-1cs424ns93, 1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, alpha-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, glucopyranose, pentaacetate, .alpha.-d, a-d-glucose pentaacetate
PubChem CID 2723636
IUPAC Name [3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 113°C
Color White
Quantity 250 g
Formula Weight 390.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Penta-O-acetyl-alpha-D-glucopyranose
EINECSNumber (8)-0053
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.