Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penta-O-acetyl-beta-D-galactopyranose 97.0+%, TCI America™
SDP

Catalog No. G0247250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0247250G 250 g
G024725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0247250G Supplier TCI America Supplier No. G0247250G

Chemical Identifiers

CAS 4163-60-4
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD00063259
InChI Key LPTITAGPBXDDGR-LYYZXLFJSA-N
Synonym beta-d-galactose pentaacetate, penta-o-acetyl-beta-d-galactopyranose, b-d-galactose pentaacetate, beta-d-galactopyranose pentaacetate, beta-d-galactosepentaacetate, beta-d-galactopyranose, pentaacetate, 2s,3r,4s,5s,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 1,2,3,4,6-penta-o-acetyl-beta-d-galactose, 1,2,3,4,6-penta-o-acetyl-beta-d-galactopyranose, 2r,3s,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate
PubChem CID 94752
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 145°C
Color White
Quantity 250 g
Formula Weight 390.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Penta-O-acetyl-beta-D-galactopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.