Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentacosanoic Acid 96.0+%, TCI America™
SDP

Catalog No. P08821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08821G 1 g
P08825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08821G Supplier TCI America Supplier No. P08821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 506-38-7
Molecular Formula C25H50O2
Molecular Weight (g/mol) 382.67
MDL Number MFCD00020551
InChI Key MWMPEAHGUXCSMY-UHFFFAOYSA-N
PubChem CID 10468
ChEBI CHEBI:39420
IUPAC Name pentacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
Formula Weight 382.67
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentacosanoic Acid
EINECSNumber (2)-0608
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.