Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecafluorooctanoic acid, MilliporeSigma™ Supelco™

Catalog No. 111019231 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9231 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9231 Supplier MilliporeSigma Supelco Supplier No. 33824100MG

Pentadecafluorooctanoic acid is a perfluoroalkyl carboxylic acid, which is primarily used as an ion pair reagent.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 335-67-1
Physical Form Neat
Linear Formula CF3(CF2)6 COOH
MDL Number MFCD00004174
Melting Point 55°C to 56°C
Boiling Point 189°C (736 mmHg)
Synonym PFOA; Perfluorocaprylic acid; Perfluorooctanoic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 414.07
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.