Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecane, Spectrum™ Chemical
SDP

Catalog No. 18621345 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
5 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-345 5 gal.
18-621-342 100 mL
18-621-343 500 mL
18-621-344 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-621-345 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P22035GLBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 629-62-9

Specifications

Boiling Point 115.5°C (239.9°F)
Packaging Drum
Quantity 5 gal.
Molecular Weight (g/mol) 80.09
Percent Purity 99%
Grade Ungraded
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Pyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.