Learn More
Cayman Chemical ANTIBODY AZD 7648 25mg
SDP
Supplier:  Cayman Chemical 2859825 MG
Encompass_Preferred
A GVIA iPLA2 inhibitor inhibits GVIA iPLA2 by 99 8 at 0 091 mole fraction selective for GVIA iPLA2 over GIVA cPLA2 and GV sPLA2 where it shows less than 25 and 32 8 inhibition respectively
Catalog No. 50-293-8599
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.