Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaerythritol Tribromide 98.0+%, TCI America™
SDP

Catalog No. T168725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T168725G 25 g
T1687500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T168725G Supplier TCI America Supplier No. T168725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1522-92-5
Molecular Formula C5H9Br3O
Molecular Weight (g/mol) 324.838
MDL Number MFCD00021982
InChI Key QEJPOEGPNIVDMK-UHFFFAOYSA-N
Synonym 3-Bromo-2,2-bis(bromomethyl)-1-propanol, Tribromoneopentyl Alcohol, 2,2,2-Tris(bromomethyl)ethanol
PubChem CID 15206
IUPAC Name 3-bromo-2,2-bis(bromomethyl)propan-1-ol
SMILES C(C(CBr)(CBr)CBr)O

Specifications

Melting Point 67°C
Color White
Boiling Point 131°C
Quantity 25 g
Formula Weight 324.84
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pentaerythritol Tribromide
EINECSNumber (2)-2002
RTECSNumber UA7410000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.