Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaethylene Glycol Monobenzyl Ether 95.0+%, TCI America™
SDP

Catalog No. P14351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14351G 1 g
P14355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14351G Supplier TCI America Supplier No. P14351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57671-28-0
Molecular Formula C17H28O6
Molecular Weight (g/mol) 328.405
MDL Number MFCD06797154
InChI Key UMUSOTNGYAALST-UHFFFAOYSA-N
Synonym 2-[2-[2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol
PubChem CID 13553692
IUPAC Name 2-[2-[2-[2-(2-phenylmethoxyethoxy)ethoxy]ethoxy]ethoxy]ethanol
SMILES C1=CC=C(C=C1)COCCOCCOCCOCCOCCO

Specifications

Color Yellow
Quantity 1 g
Formula Weight 328.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentaethylene Glycol Monobenzyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.