Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzyl Alcohol 96.0+%, TCI America™
SDP

Catalog No. P084925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P084925G 25 g
P08495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P084925G Supplier TCI America Supplier No. P084925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 440-60-8
Molecular Formula C7H3F5O
Molecular Weight (g/mol) 198.092
MDL Number MFCD00004602
InChI Key PGJYYCIOYBZTPU-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzyl alcohol, pentafluorobenzyl alcohol, perfluorophenyl methanol, pentafluorophenyl methanol, 2,3,4,5,6-pentafluorophenyl methanol, 2,3,4,5,6-pentafluorobenzenemethanol, hydroxymethyl pentafluorobenzene, benzenemethanol, 2,3,4,5,6-pentafluoro, pentafluorobenzylalcohol, 2,3,4,5,6-pentafluorobenzylalcohol
PubChem CID 9923
ChEBI CHEBI:44903
IUPAC Name (2,3,4,5,6-pentafluorophenyl)methanol
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)O

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 182°C
Quantity 25 g
Formula Weight 198.09
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Pentafluorobenzyl Alcohol
RTECSNumber DP0695000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.