Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent
GSA_VA

Catalog No. PI58220 Shop All Thermo Scientific Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI58220 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI58220 Supplier Thermo Scientific™ Supplier No. TS58220
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Derivatize carboxyl acids, phenols, and sulfonamides for electron-capture detection (ECD) GC analysis with the Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent.

Derivatize carboxyl acids, phenols, and sulfonamides for electron-capture detection (ECD) GC analysis with the Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent. Reaction times are fast (∼20 minutes). Derivatives are highly EC-sensitive, making them useful in low-level determinations of fatty acids. PFBBr has been applied in analyzing trace organics in asphalts and as a fingerprinting technique for identifying asphalt pollutants found in surface waters.

  • Fast reaction times for extraction alkylation technique: ∽20 minutes
  • Derivatives are highly EC-sensitive, making them useful in low-level determinations of fatty acids
  • Analysis of trace organics in asphalt

Recommended for:

Analysis of trace organics in asphalt

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Hazards Corrosive
Standard Components alpha-Bromo-2,3,4,5,6-pentafluorotoluene
Standard Concentration 98 to 100%
Quantity 5 g
Storage Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.