Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoroiodobenzene (stabilized with Copper chip) 99.0+%, TCI America™
SDP

Catalog No. P118825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P118825G 25 g
P11885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P118825G Supplier TCI America Supplier No. P118825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 827-15-6
Molecular Formula C6F5I
Molecular Weight (g/mol) 293.962
MDL Number MFCD00001032
InChI Key OPYHNLNYCRZOGY-UHFFFAOYSA-N
Synonym iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982
PubChem CID 70008
IUPAC Name 1,2,3,4,5-pentafluoro-6-iodobenzene
SMILES C1(=C(C(=C(C(=C1F)F)I)F)F)F

Specifications

Color Yellow
Boiling Point 162°C
Quantity 25 g
Formula Weight 293.96
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluoroiodobenzene (stabilized with Copper chip)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.