Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenyl 4-[5-(trifluoromethyl)pyrid-2-yl]benzoate, ≥95%, Thermo Scientific™

Catalog No. CC53026CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC53026CB 250 mg Amber Glass Bottle
CC53026DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC53026CB Supplier Thermo Scientific Chemicals Supplier No. CC53026CB

Chemical Identifiers

CAS 910036-89-4
Molecular Formula C19H7F8NO2
Molecular Weight (g/mol) 433.257
MDL Number MFCD09817480
InChI Key FXVFNQGTIWSDPI-UHFFFAOYSA-N
Synonym pentafluorophenyl 4-5-trifluoromethyl pyridin-2-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-5-trifluoromethyl pyridin-2-yl benzoate, pentafluorophenyl 4-5-trifluoromethyl pyrid-2-yl benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-5-trifluoromethyl pyridin-2-yl benzoate, 4-5-trifluoromethyl-2-pyridinyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229536
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 4-[5-(trifluoromethyl)pyridin-2-yl]benzoate
SMILES C1=CC(=CC=C1C2=NC=C(C=C2)C(F)(F)F)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 433.25
Percent Purity 90%
Chemical Name or Material Pentafluorophenyl 4-[5-(trifluoromethyl)pyrid-2-yl]benzoate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.