Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenylhydrazine, 97%, Thermo Scientific Chemicals

Catalog No. AAA1330206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1330206 5 g
AAA1330214 25 g
AAA1330222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1330206 Supplier Thermo Scientific Chemicals Supplier No. A1330206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 828-73-9
Molecular Formula C6H3F5N2
Molecular Weight (g/mol) 198.096
MDL Number MFCD00007574
InChI Key BYCUWCJUPSUFBX-UHFFFAOYSA-N
Synonym pentafluorophenylhydrazine, pentafluorophenyl hydrazine, perfluorophenyl hydrazine, hydrazine, pentafluorophenyl, 2,3,4,5,6-pentafluorophenyl hydrazine, 1-2,3,4,5,6-pentafluorophenyl hydrazine, pubchem3316, pentafluorophenyl-hydrazine
PubChem CID 13236
IUPAC Name (2,3,4,5,6-pentafluorophenyl)hydrazine
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)NN

Specifications

Melting Point 73°C to 78°C
Quantity 5 g
Beilstein 747800
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 198.09
Percent Purity 97%
Chemical Name or Material Pentafluorophenylhydrazine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-586-7
RTECSNumber MV8325000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.