Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane, ≥99.0%, Solstice™

Catalog No. 60047539 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-539 2.5 L Poly Bottle
60-047-538 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-539 Supplier Solstice Supplier No. 768692.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, for pesticide residue analysis,≥99.0%

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

pH 5.7 (at 22°C)
Melting Point 750°C
Density 0.626g/mL at 25°C
Quantity 2.5 L
Explosion Limit 8.3%
Ignition Point 500°C
Packaging Poly Bottle
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Boiling Point 1393°C (1013 hPa)
Solubility Information 946g/L at 25°C
Formula Weight 72.15g/mol
Percent Purity ≥99.0%
Chemical Name or Material Pentane
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331
DOTInformation class 3 - PG 2 - Pentanes
RTECSNumber 203-692-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.