Learn More
Sigma Aldrich Fine Chemicals Biosciences Pentane anhydrous 99
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 236705
Encompass
Pentane, a linear aliphatic hydrocarbon, is one of the main constituents of light straight-run (LSR) gasoline fraction in crude oils. Its isomerization can improve the octane number of gasoline. This has led to the development of a number of metal catalysts, which can effectively isomerize n-pentane to iso-pentane.
Catalog No. 50-290-6740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.