Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane Anhydrous, DriSolv™, MilliporeSigma™

Catalog No. MPX01716 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX01716 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX01716 Supplier MilliporeSigma Supplier No. PX01716
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Septum-Sealed Bottle

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -129.6°C
Color Colorless
Quantity 1 L
Flash Point Closed cup: <-40°C (<-40°F)
Ignition Point 260°C (500°F)
Odor Gas
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 59 kPa (442.83mm Hg) [20°C]
UN Number 1265
Boiling Point 36.06°C
Solubility Information Insoluble in water
Formula Weight 72.15
Percent Purity ≥99%
Chemical Name or Material Pentane Anhydrous
Show More Show Less
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept uprightto prevent leakage.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.