Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane, Reagent grade, 98%,Solstice

Catalog No. 60044920 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
6 x 1 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-920 6 x 1 L Glass Bottle
60-044-914 1 L Glass Bottle
60-044-915 2.5 L Glass Bottle
60-044-919 5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-044-920 Supplier Solstice Supplier No. 1589416X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -131°C
Color Colorless
Density 0.626g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.083
Flash Point −49°C (−56.2°F)
Ignition Point 500°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.358(lit.)
Linear Formula CH3(CH2)3CH3
Vapor Pressure 26.85psi (55°C), 8.43psi (20°C)
UN Number UN1265
Beilstein 969132
Boiling Point 37°C
Formula Weight 72.15g/mol
Percent Purity 98%
Grade Reagent
Chemical Name or Material Pentane
Show More Show Less
Hazard Statement P210-P301 + P310-P331-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H336-H411
DOTInformation Class 3 - PG 2 - Pentanes
EINECSNumber 203-692-4
RTECSNumber RZ9450000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.