Learn More
Sigma Aldrich Fine Chemicals Biosciences Pentane suitable for HPLC
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 34956
Encompass
Pentane (n-pentane) is a hydrocarbon. Its hydroisomerization using a nanocrystalline beta (BEA) zeolite has been proposed. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid.
Catalog No. 50-290-6729