Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentatriacontane 95.0+%, TCI America™
SDP

Catalog No. P006210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P006210G 10 g
P00621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P006210G Supplier TCI America Supplier No. P006210G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 630-07-9
Molecular Formula C35H72
Molecular Weight (g/mol) 492.961
MDL Number MFCD00059397
InChI Key VHQQPFLOGSTQPC-UHFFFAOYSA-N
Synonym n-pentatriacontane, 8ci 9ci, acmc-209nc1, pentatriacontane, analytical standard
PubChem CID 12413
IUPAC Name pentatriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 74°C
Color White
Boiling Point 331°C
Quantity 10 g
Formula Weight 492.96
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentatriacontane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.