Learn More
Medchemexpress LLC HY-A0203A 100mg Medchemexpress, Pentosan Polysulfate (Sodium) CAS:140207-93-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0203A100MG
Encompass_Preferred
Medchemexpress, HY-A0203A 100mg Pentosan Polysulfate (Sodium) CAS:140207-93-8 Pentosan Polysulfate Sodium is an orally bioavailable, semi-synthetic medication with anti-inflammatory and pro-chondrogenic properties. Pentosan Polysulfate Sodium also is a potent and selective anti-HIV agent. Pentosan Polysulfate Sodium is used for the treatment of interstitial cystitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2335
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.