Learn More
Enzo Life Sciences Pentoxifylline (5g). CAS: 6493-05-6
SDP
Supplier:  Enzo Life Sciences ALX270112G005
Encompass_Preferred
Phosphodiesterase inhibitor. Blocks the synthesis of TNF-α. Improves blood flow by decreasing blood viscosity. Alternative name: Trental, 3,7-Dihydro-3,7-dimethyl-1-(5-oxohexyl)-1H-purine-2,6-dione. Formula: C13H18N4O3. MW: 278.3. Purity: ≥99% (Titration). Solubility: Soluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-8412
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.