Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentraxin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7205982 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9013 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9013 Supplier Proteintech Group Inc Supplier No. 137971AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of "long pentraxins", which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF. Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.

Specifications

Antigen Pentraxin 3
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PTX3
Gene Accession No. P26022, P48759
Gene Alias Pentaxin related protein PTX3, Pentraxin 3, Pentraxin related protein PTX3, PTX3, TNF alpha induced protein 5, TNFAIP5, TSG 14, TSG14
Gene Symbols PTX3
Host Species Rabbit
Immunogen Pentraxin 3 Fusion Protein Ag4624
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19288, 5806, 689388
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.