Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P226725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2267-25G 25 g
P22675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2267-25G Supplier TCI America Supplier No. P226725G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4737-50-2
Molecular Formula C5H13BO2
Molecular Weight (g/mol) 115.967
MDL Number MFCD01074651
InChI Key ABWPXVJNCQKYDR-UHFFFAOYSA-N
Synonym 1-Pentaneboronic Acid
PubChem CID 352103
IUPAC Name pentylboronic acid
SMILES B(CCCCC)(O)O

Specifications

Melting Point 94°C
Color White
Quantity 25 g
Formula Weight 115.97
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.