Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentyltriethoxysilane 97.0+%, TCI America™
SDP

Catalog No. P073625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P073625ML 25 mL
P07365ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P073625ML Supplier TCI America Supplier No. P073625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2761-24-2
Molecular Formula C11H26O3Si
Molecular Weight (g/mol) 234.411
MDL Number MFCD00026757
InChI Key FHVAUDREWWXPRW-UHFFFAOYSA-N
Synonym Amyltriethoxysilane, Triethoxy(pentyl)silane
PubChem CID 17695
IUPAC Name triethoxy(pentyl)silane
SMILES CCCCC[Si](OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 205°C
Quantity 25 mL
Formula Weight 234.41
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentyltriethoxysilane
EINECSNumber (2)-2052&(2)-2053
RTECSNumber VV4750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.