Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings

Catalog No. 03255019 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Cartridge Holder Kit
Tubing
Includes:
Cartridge Holder, Tubing, Fittings
Tubing, Fittings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
03-255-019 Tubing Tubing, Fittings
03-255-020 Cartridge Holder Kit Cartridge Holder, Tubing, Fittings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-255-019 Supplier Thermo Scientific™ Supplier No. 174501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap™ Neo Trap Cartridge to your HPLC system.

Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap Neo Trap Cartridge (Catalog Number 174500) to your HPLC system.

See Viper and nanoViper resources

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 30 μm
For Use With PepMap™ Neo Trap Cartridge (part number 174500)
Includes Tubing, Fittings
Length (Metric) 100 mm
Max. Pressure 22,000 psi (1500 bar)
Temperature 60°C
Product Line PepMap Neo
Material PEEK
Description PEEK Tubing with nanoViper™ Fittings
For Use With (Application) Trap and Elute, Peptide Mapping
Product Type Tubing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.