Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peppermint Oil, NF, 5%, Spectrum™ Chemical
SDP

Catalog No. 18605012 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
10 kg
180 kg
Packaging:
Steel Drum
Steel Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-012 10 kg Steel Pail
18-605-013 180 kg Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-605-012 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PE10510KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

PE105, 8006-90-4

Spectrum™ Chemical Peppermint Oil, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8006-90-4

Specifications

CAS Max % 1
Packaging Steel Pail
Refractive Index 1.459-1.465
Quantity 10 kg
Percent Purity 5%
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.