Learn More
Mabtech Inc PepPool CyCMV+RhCMV (TP-UL83+Rh112), peptide pool, 266 peptides total, 15-mers, mean purity 83%, lyophilized, two vials, 25 μg per peptide, intended for immune response studies
Supplier:  Mabtech Inc 36921
Encompass
PepPool CyCMV+RhCMV (TP-UL83+Rh112), peptide pool, 266 peptides total, 15-mers, mean purity 83%, lyophilized, two vials, 25 μg per peptide, intended for immune response studies
Catalog No. NC2598644
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.