Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ PeproGMP® Human Flt-3 Ligand (FLT3L) Recombinant Protein, PeproTech®

Catalog No. 502558471
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-8471 1 mg
50-255-8472 100 μg
01-672-637 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-255-8471 Supplier Gibco™ Supplier No. GMP300191MG

Request a quote on this item.

Recombinant Protein

Multiple isoforms of Flt3-Ligand have been identified. The predominant biologically active form is anchored to the cell surface as the extracellular domain of a transmembrane protein (209 a.a.). The membrane-bound isoform can be proteolytically cleaved to generate a biologically active soluble isoform. Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49771
For Use With (Application) Control, Western Blot Control, Bioactivity
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 2323
Molecular Weight (g/mol) 17.6 kDa
Name PeproGMP™ Human Flt-3 Ligand (FLT3L)
Purification Method Purified
Quantity 1 mg
Storage Requirements -20°C or -80°C if preferred
Regulatory Status GMP
Endotoxin Concentration ≤0.1 EU/μg
Gene Alias FL; Flt 3 ligand; Flt3; Flt3 L; Flt3 ligand; Flt3l; Flt3lg; fms related receptor tyrosine kinase 3 ligand; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; fms-related tyrosine kinase 3; fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine
Common Name FLT3LG
Gene Symbol FLT3LG
Biological Activity Determined by its ability to stimulate human AML5 cell proliferation. PeproGMP Recombinant Human Flt3-Ligand has been tested against the WHO standard NIBSC 96/532.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MTQDCSFQHS PISSDFAVKI RELSDYLLQD YPVTVASNLQ DEELCGGLWR LVLAQRWMER LKTVAGSKMQ GLLERVNTEI HFVTKCAFQP PPSCLRFVQT NISRLLQETS EQLVALKPWI TRQNFSRCLE LQCQPDSSTL PPPWSPRPLE ATAPTA
Content And Storage -20°Cor to +80°C if preferred
Activity Determined by its ability to stimulate human AML5 cell proliferation. PeproGMP Recombinant Human Flt3-Ligand has been tested against the WHO standard NIBSC 96/532. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Expression System E. coli
Form Lyophilized
Product Line PeproGMP
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.