Learn More
Description
- Material: Pepsin Powder
- Color: White to yellow
- Quantity: 100 grams
- Packaging: Powder form
- Usage: For educational use only – no Certificate of Analysis (C of A) available
- Ideal For: Enzyme activity studies, protein digestion experiments, biology and chemistry classrooms
This product is for educational use - no C of A is available.
Specifications
Specifications
| Color | White to Yellow |
| Physical Form | Flakes or Powder |
| Quantity | 30 g |
| Packaging | Powder |
| Chemical Name or Material | Pepsin Powder |
Safety and Handling
| ShelfLife | 5 years |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
